Learn More
Medchemexpress LLC α-NAD (α-Nicotinamide Adenine Dinucleotide) | 7298-93-3 | 98.0% | 5 MG
SDP
Catalog No. 5000406772
Encompass_Preferred
α-NAD (α-Nicotinamide Adenine Dinucleotide) serves as a substrate for renalase and functions as an analog of β-NAD, influencing the kinetics of NAD-dependent enzymes, while also playing a crucial role as a cofactor in mitochondrial oxidation-reduction reactions.
  • Functions as a substrate for renalase.
  • Acts as an analog of β-NAD.
  • Impacts the kinetics of NAD-dependent enzymes.
  • Plays a vital role as a cofactor in mitochondrial oxidation-reduction reactions.
Catalog No. 50-004-06772 Supplier Medchemexpress LLC Supplier No. HYB0445B5MG
α-NAD (α-Nicotinamide Adenine Dinucleotide) serves as a substrate for renalase and functions as an analog of β-NAD, influencing the kinetics of NAD-dependent enzymes, while also playing a crucial role as a cofactor in mitochondrial oxidation-reduction reactions.
  • Functions as a substrate for renalase.
  • Acts as an analog of β-NAD.
  • Impacts the kinetics of NAD-dependent enzymes.
  • Plays a vital role as a cofactor in mitochondrial oxidation-reduction reactions.