Learn More
Medchemexpress LLC NAD(P)H-Nitrate reductase | 9029-27-0 | ≥0.5 U/mg solid | 180 kDa (gel filtration) | 1 U
SDP
Catalog No. 5000398649
Encompass_Preferred
NAD(P)H-Nitrate reductase is isolated from Aspergillus niger and catalyzes the reduction of nitrate to nitrite via a two-electron transfer. In plants, the electron donor for Nitrate reductase is NADPH or NADH:Nitrate reductase, and it can also be a bispecific NAD(P)H: Nitrate reductase. This product is for research use only and is not sold to patients.
  • Isolated from Aspergillus niger.
  • Catalyzes the reduction of nitrate to nitrite via a two-electron transfer.
  • Research use only.
Catalog No. 50-003-98649 Supplier Medchemexpress LLC Supplier No. HYP29961U
NAD(P)H-Nitrate reductase is isolated from Aspergillus niger and catalyzes the reduction of nitrate to nitrite via a two-electron transfer. In plants, the electron donor for Nitrate reductase is NADPH or NADH:Nitrate reductase, and it can also be a bispecific NAD(P)H: Nitrate reductase. This product is for research use only and is not sold to patients.
  • Isolated from Aspergillus niger.
  • Catalyzes the reduction of nitrate to nitrite via a two-electron transfer.
  • Research use only.