Learn More
Bioworld NAD-SEPAROPOREAGAROSE) 4B
Catalog No. 50197721
Encompass
NAD-SEPAROPORE (AGAROSE) 4B, LYOPHILIZED: NAD-Separopore NAD covalently bound to Separoporehas a high affinity for NAD- dependent enzymes and dehydrogenases. NAD Separopore has been used to purify Vibrio cholerae toxin, diptheria toxin ...
Catalog No. 50-197-721 Supplier Bioworld Supplier No. 201810242
NAD-SEPAROPORE (AGAROSE) 4B, LYOPHILIZED: NAD-Separopore NAD covalently bound to Separoporehas a high affinity for NAD- dependent enzymes and dehydrogenases. NAD Separopore has been used to purify Vibrio cholerae toxin, diptheria toxin ...