Learn More
Bioworld NAD-SEPAROPOREAGAROSE) 4B
Supplier:  Bioworld 201810241
Encompass
NAD-SEPAROPORE (AGAROSE) 4B, LYOPHILIZED: NAD-Separopore NAD covalently bound to Separoporehas a high affinity for NAD- dependent enzymes and dehydrogenases. NAD Separopore has been used to purify Vibrio cholerae toxin, diptheria toxin ...
Catalog No. 50-197-720