Learn More
Enzo Life Sciences NAD (SIRT Substrate) (500µl)
SDP
Catalog No. 502011111
Encompass_Preferred
Formulation: 50mM Nicotinamide adenine dinucleotide (oxidized form) in 50mM Tris/HCl, pH 8.0, containing 137mM sodium chloride, 2.7mM KCl and 1mM MgCl2. Long Term Storage: -80°C.
Catalog No. 50-201-1111 Supplier Enzo Life Sciences Supplier No. BMLKI2820500
Formulation: 50mM Nicotinamide adenine dinucleotide (oxidized form) in 50mM Tris/HCl, pH 8.0, containing 137mM sodium chloride, 2.7mM KCl and 1mM MgCl2. Long Term Storage: -80°C.