Learn More
Cayman Chemical -NAD sodIum salt 6.9mg
SDP
Supplier:  Cayman Chemical 383856.9 MG
Encompass_Preferred
A fluorescent probe for NAD+-dependent enzymes; ex/em = 300/410 nm, respectively
Catalog No. 50-291-7191