Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp NADH [B-Nicotinamide adenine dinucleotide, reduced form, disodium salt], 1 Gram
SDP

Catalog No. 50213547
Encompass_Preferred

Substance Name: B-Nicotinamide adenine dinucleotide, reduced form, disodium salt
CAS Number: 606-68-8
Molecular Formula: C21H27N7O14 P2Na2 •: 3H2O
Molecular Weight: 763.5
Chemical Purity: >98%
Soluble in: Water
Store at: 2-8°C

Catalog No. 50-213-547 Supplier Research Products International Corp Supplier No. N201001.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Substance Name: B-Nicotinamide adenine dinucleotide, reduced form, disodium salt
CAS Number: 606-68-8
Molecular Formula: C21H27N7O14 P2Na2 •: 3H2O
Molecular Weight: 763.5
Chemical Purity: >98%
Soluble in: Water
Store at: 2-8°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.