Learn More
Research Products International Corp NADH [B-Nicotinamide adenine dinucleotide, reduced form, disodium salt], 5 Grams

Supplier: Research Products International Corp N201005.0
Substance Name: B-Nicotinamide adenine dinucleotide, reduced form, disodium salt
CAS Number: 606-68-8
Molecular Formula: C21H27N7O14 P2Na2 •: 3H2O
Molecular Weight: 763.5
Chemical Purity: >98%
Soluble in: Water
Store at: 2-8°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.