Learn More
Gold Biotechnology Inc NADH Beta Reduced Sodium Salt
SDP
Supplier:  Gold Biotechnology Inc N0351
Encompass_Preferred
Reduced -nicotinamide adenine dinucleotide (NADH) plays a major role in metabolism as a cofactor in redox reactions and as a mobile electron carrier NADH is a high energy compound that donates electrons to the electron transport chain to provide energy for ATP production by oxidative phosphorylation NADH is a required oxidizing cosubstrate in fermentation which regenerates NADH is fluorescent which provides for a relatively simple way to detect NADH in biological samples NADH is also used in enzyme cycling assays to detect relevant biological molecules in tissues
Catalog No. 50-005-62150