Learn More
Medchemexpress LLC HY-F0001 100mg Medchemexpress, NADH (disodium salt) CAS:606-68-8 Purity:>98%
SDP
Catalog No. 501933237
Encompass_Preferred
Medchemexpress, HY-F0001 100mg NADH (disodium salt) CAS:606-68-8 NADH disodium salt is a coenzyme of a large number of oxidoreductases. NADH is a coenzyme that functions as a regenerating electron donor in catabolic processes including glycolysis, beta-oxidation and the citric acid cycle. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3237 Supplier Medchemexpress LLC Supplier No. HYF0001100MG
Medchemexpress, HY-F0001 100mg NADH (disodium salt) CAS:606-68-8 NADH disodium salt is a coenzyme of a large number of oxidoreductases. NADH is a coenzyme that functions as a regenerating electron donor in catabolic processes including glycolysis, beta-oxidation and the citric acid cycle. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.