Learn More
Selleck Chemical LLC NADH Disodium Salt Hydrate
SDP
Supplier:  Selleck Chemical LLC S634925MG
Encompass_Preferred
NADH is a coenzyme that functions as a regenerating electron donor in catabolic processes including glycolysis -oxidation and the citric acid cycle
Catalog No. 50-000-04010