Learn More
Abcam NADH disodium salt trihydrate, Coenzyme, 1G
Catalog No. 502704733
Encompass_Preferred
MW 763.4 g/mol, Purity >95%. Sodium salt of NADH, a coenzyme that functions as a regenerating electron donor in catabolic processes including glycolysis, β-oxidation and the citric acid cycle (Krebs cycle, TCA cycle). Also participates in cell signaling events, for example as a substrate for the poly (ADP-ribose) polymerases (PARPs) during the DNA damage response.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4733 Supplier Abcam Supplier No. AB1463151G
MW 763.4 g/mol, Purity >95%. Sodium salt of NADH, a coenzyme that functions as a regenerating electron donor in catabolic processes including glycolysis, β-oxidation and the citric acid cycle (Krebs cycle, TCA cycle). Also participates in cell signaling events, for example as a substrate for the poly (ADP-ribose) polymerases (PARPs) during the DNA damage response.
The product is subject to the following: Abcam Restricted Use Statement