Learn More
Medchemexpress LLC NADH-IN-1 | 1432445-15-2 | MFCD07109669 | 99.4% | 382.44 g·mol⁻¹ | C19H21F3N2OS | 5 MG
SDP
Catalog No. 5000222290
Encompass_Preferred
NADH-IN-1 is a small-molecule research inhibitor of NADH:ubiquinone oxidoreductase (complex I) supplied for laboratory use. It is provided as a high-purity solid in multiple milligram quantities and as a 10 mM solution in DMSO. CAS: 1432445-15-2; formula: C19H21F3N2OS.
Catalog No. 50-002-22290 Supplier Medchemexpress LLC Supplier No. HY1460365MG
NADH-IN-1 is a small-molecule research inhibitor of NADH:ubiquinone oxidoreductase (complex I) supplied for laboratory use. It is provided as a high-purity solid in multiple milligram quantities and as a 10 mM solution in DMSO. CAS: 1432445-15-2; formula: C19H21F3N2OS.