Learn More
TRIBIOSCIENCE INC NADH/NAD Quantification Colorimetric Assay Kit, 100 Tests
SDP
Catalog No. NC2886948
Encompass_Preferred
NADH/NAD Quantification Colorimetric Assay provides the easiest and most accurate approach to measuring NADH from a variety of samples, which can be measured at OD 460 nm.
Catalog No. NC2886948 Supplier TRIBIOSCIENCE INC Supplier No. TBS2029
NADH/NAD Quantification Colorimetric Assay provides the easiest and most accurate approach to measuring NADH from a variety of samples, which can be measured at OD 460 nm.