Learn More
Apexbio Technology LLC NADH (sodium salt)(Synonyms: Reduced β-Nicotinamide adenine dinucleotide disodium salt, NADH disodium salt), 1g, CAS: 606-68-8.
SDP
Catalog No. 5000061357
Encompass_Preferred
NADH (sodium salt) (CAS 606-68-8) is the reduced form of nicotinamide adenine dinucleotide functioning as an essential coenzyme involved in cellular bioenergetics Biologically NADH participates as an electron carrier in metabolic pathways including glycolysis -oxidation and the tricarboxylic acid (TCA) cycle facilitating ATP generation through electron transport In biomedical research NADH (sodium salt) is frequently applied to investigate cellular energy metabolism and elucidate mechanisms underpinning metabolic disorders
Catalog No. 50-000-61357 Supplier Apexbio Technology LLC Supplier No. C44951000
NADH (sodium salt) (CAS 606-68-8) is the reduced form of nicotinamide adenine dinucleotide functioning as an essential coenzyme involved in cellular bioenergetics Biologically NADH participates as an electron carrier in metabolic pathways including glycolysis -oxidation and the tricarboxylic acid (TCA) cycle facilitating ATP generation through electron transport In biomedical research NADH (sodium salt) is frequently applied to investigate cellular energy metabolism and elucidate mechanisms underpinning metabolic disorders