Learn More
AdipoGen NADK h Hishighly active
SDP
Catalog No. 502562772
Encompass_Preferred
Protein. Human full length NAD kinase aa 1-446 is fused at the N-terminus to a His-tag. Source E. coli. Endotoxin content N/A. Free of any nucleotide converting side-activities. Liquid. Ammonium sulphate suspension. Purity >95% SDS-PAGE. NAD kinase catalyzes the transfer of a phosphate group from ATP to NAD to generate NADP, which in its reduced form acts as an electron donor for biosynthetic reactions. NADP is an essential coenzyme in metabolism and provides reducing power to biosynthetic processes such as fatty acid biosynthesis.
Catalog No. 50-256-2772 Supplier AdipoGen Supplier No. AG40T0091C050
Protein. Human full length NAD kinase aa 1-446 is fused at the N-terminus to a His-tag. Source E. coli. Endotoxin content N/A. Free of any nucleotide converting side-activities. Liquid. Ammonium sulphate suspension. Purity >95% SDS-PAGE. NAD kinase catalyzes the transfer of a phosphate group from ATP to NAD to generate NADP, which in its reduced form acts as an electron donor for biosynthetic reactions. NADP is an essential coenzyme in metabolism and provides reducing power to biosynthetic processes such as fatty acid biosynthesis.