Learn More
Cayman Chemical NAD/NADH Cell-Based Assay Kit, 1 ea
SDP
Catalog No. 502897048
Encompass_Preferred
Nicotinamide adenine dinucleotide (NAD) exists in an oxidized form, NAD+, as well as a reduced form, NADH. NAD, the main free form in cells, functions in modulating cellular redox status and by controlling signaling and transcriptional events, making it an important cofactor when investigating normal cellular function. Cayman’s NAD/NADH Cell-Based Assay Kit provides a colorimetric ethod for measuring intracellular NAD+and NADH in cultured cells. In this assay, NAD+ found in cell samples is reduced to NADH by alcohol dehydrogenase during the oxidation of ethanol to acetaldehyde. The newly formed and the existing NADH found in the samples is then oxidized resulting in the reduction of a tetrazolium salt substrate (WST-1) to a highly-colored formazan which absorbs at 450 nm. The amount of formazan produced is proportional to the amount of total NAD in the cell lysate and can be used as an indicator of the total cellular NAD concentration.
Catalog No. 50-289-7048 Supplier Cayman Chemical Supplier No. 6004801 EA
Nicotinamide adenine dinucleotide (NAD) exists in an oxidized form, NAD+, as well as a reduced form, NADH. NAD, the main free form in cells, functions in modulating cellular redox status and by controlling signaling and transcriptional events, making it an important cofactor when investigating normal cellular function. Cayman’s NAD/NADH Cell-Based Assay Kit provides a colorimetric ethod for measuring intracellular NAD+and NADH in cultured cells. In this assay, NAD+ found in cell samples is reduced to NADH by alcohol dehydrogenase during the oxidation of ethanol to acetaldehyde. The newly formed and the existing NADH found in the samples is then oxidized resulting in the reduction of a tetrazolium salt substrate (WST-1) to a highly-colored formazan which absorbs at 450 nm. The amount of formazan produced is proportional to the amount of total NAD in the cell lysate and can be used as an indicator of the total cellular NAD concentration.