Learn More
AdipoGen Nadolol
SDP
Catalog No. 502560395
Encompass_Preferred
Chemical. CAS 42200-33-9. Formula C17H27NO4. MW 309.4. Nadolol is a polar and hydrophilic, non-selective beta-adrenergic antagonist with antihypertensive and antiarrhythmic activities. It is a long-acting beta-blocker, which displays activity against beta-1 and beta-2 receptors. It has a preference for beta-1 receptors, which are predominantly located in the heart, thereby inhibiting the effects of catecholamines and causing a decrease in heart rate and blood pressure. Nadolols inhibition of beta-1 receptors in the heart and kidney leads to its effects on lowering blood pressure. Its inhibition of beta-2 receptors, which are mainly located in the bronchial smooth muscle of the airways, leads to airway constriction similar to that seen in asthma.
Catalog No. 50-256-0395 Supplier AdipoGen Supplier No. CDXN0141G001
Chemical. CAS 42200-33-9. Formula C17H27NO4. MW 309.4. Nadolol is a polar and hydrophilic, non-selective beta-adrenergic antagonist with antihypertensive and antiarrhythmic activities. It is a long-acting beta-blocker, which displays activity against beta-1 and beta-2 receptors. It has a preference for beta-1 receptors, which are predominantly located in the heart, thereby inhibiting the effects of catecholamines and causing a decrease in heart rate and blood pressure. Nadolols inhibition of beta-1 receptors in the heart and kidney leads to its effects on lowering blood pressure. Its inhibition of beta-2 receptors, which are mainly located in the bronchial smooth muscle of the airways, leads to airway constriction similar to that seen in asthma.