Learn More
Medchemexpress LLC Nadolol-d9 | 94513-92-5 | MFCD00049405 | 97.0% | 5 MG
SDP
Catalog No. 5000408789
Encompass_Preferred
Nadolol-d9 is a deuterium labeled Nadolol, functioning as a non-selective and orally active β-adrenergic receptors blocker. It is intended for research use only.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Supports NMR, GC-MS, or LC-MS applications
Catalog No. 50-004-08789 Supplier Medchemexpress LLC Supplier No. HYB0804S5MG
Nadolol-d9 is a deuterium labeled Nadolol, functioning as a non-selective and orally active β-adrenergic receptors blocker. It is intended for research use only.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Supports NMR, GC-MS, or LC-MS applications