Learn More
Medchemexpress LLC Nadolol-d9 (SQ-11725-d9) | 94513-92-5 | 97.0% | 1 MG
SDP
Catalog No. 5000408086
Encompass_Preferred
Nadolol-d9 is the deuterium labeled Nadolol. Nadolol is a non-selective and orally active β-adrenergic receptors blocker. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-08086 Supplier Medchemexpress LLC Supplier No. HYB0804S1MG
Nadolol-d9 is the deuterium labeled Nadolol. Nadolol is a non-selective and orally active β-adrenergic receptors blocker. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.