Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nadolol-d9 (SQ-11725-d9) | 94513-92-5 | 97.0% | 318.46 | C17H18D9NO4 | 10 MG
SDP

Catalog No. 5000297728
Encompass_Preferred

Nadolol-d9 is the deuterium-labeled analog of nadolol, a non-selective, orally active β-adrenergic receptor blocker used for analytical and research applications as a tracer or internal standard.

  • Deuterium-labeled internal standard for quantitative analysis by NMR, GC-MS, and LC-MS.
  • Non-selective β-adrenergic receptor blocker analog suitable for tracer and pharmacokinetic studies.
  • High purity (97.0%) for reliable analytical performance.
  • Molecular formula C17H18D9NO4 and molecular weight 318.46 g/mol.
  • Off-white to light yellow solid appearance.
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Available in small analytical quantities, including 10 mg packs.

Catalog No. 50-002-97728 Supplier Medchemexpress LLC Supplier No. HYB0804S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nadolol-d9 is the deuterium-labeled analog of nadolol, a non-selective, orally active β-adrenergic receptor blocker used for analytical and research applications as a tracer or internal standard.

  • Deuterium-labeled internal standard for quantitative analysis by NMR, GC-MS, and LC-MS.
  • Non-selective β-adrenergic receptor blocker analog suitable for tracer and pharmacokinetic studies.
  • High purity (97.0%) for reliable analytical performance.
  • Molecular formula C17H18D9NO4 and molecular weight 318.46 g/mol.
  • Off-white to light yellow solid appearance.
  • Storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Available in small analytical quantities, including 10 mg packs.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.