Learn More
Selleck Chemical LLC NADP E7274-50MG
SDP
Catalog No. 5000041900
Encompass_Preferred
NADP is a coenzyme involved in cellular electron transfer reactions in biological metabolism which is alternately oxidized (NADP ) and reduced (NADPH) and can maintain cellular redox homeostasis and regulate many biological events including cellular metabolism NADPH is a universal electron donor that provides reducing ability for synthetic metabolic reactions and redox balance NADPH plays a multifunctional role in regulating inflammation redox homeostasis and synthetic metabolism processes
Catalog No. 50-000-41900 Supplier Selleck Chemical LLC Supplier No. E727450MG
NADP is a coenzyme involved in cellular electron transfer reactions in biological metabolism which is alternately oxidized (NADP ) and reduced (NADPH) and can maintain cellular redox homeostasis and regulate many biological events including cellular metabolism NADPH is a universal electron donor that provides reducing ability for synthetic metabolic reactions and redox balance NADPH plays a multifunctional role in regulating inflammation redox homeostasis and synthetic metabolism processes