Learn More
Abcam NADP, Coenzyme, 1G
Catalog No. 502704731
Encompass_Preferred
MW 787.37 g/mol, Purity >=93%. NADP and NADPH form a redox pair. NADP is the oxidised form of NADPH. NADP/NADPH is a coenzyme that supports redox reactions via the transport of electrons in a vast array of applications. Acts as a coenzyme couple in various cytochrome P450 systems and oxidase/reductase reaction systems.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4731 Supplier Abcam Supplier No. AB1463161G
MW 787.37 g/mol, Purity >=93%. NADP and NADPH form a redox pair. NADP is the oxidised form of NADPH. NADP/NADPH is a coenzyme that supports redox reactions via the transport of electrons in a vast array of applications. Acts as a coenzyme couple in various cytochrome P450 systems and oxidase/reductase reaction systems.
The product is subject to the following: Abcam Restricted Use Statement