Learn More
Selleck Chemical LLC NADP disodium salt
SDP
Catalog No. 5000004813
Encompass_Preferred
Triphosphopyridine nucleotide (NADP) serves as an electron carrier in a number of reactions being alternately oxidized (NADP ) and reduced (NADPH)
Catalog No. 50-000-04813 Supplier Selleck Chemical LLC Supplier No. S64801G
Triphosphopyridine nucleotide (NADP) serves as an electron carrier in a number of reactions being alternately oxidized (NADP ) and reduced (NADPH)