Learn More
Enzo Life Sciences NADP. disodium salt (1g). CAS: 24292-60-2
SDP
Catalog No. 502009300
Encompass_Preferred
Alternative name: beta-Nicotinamide Adenine Dinucleotide Phosphate, beta-NADP, Coenzyme II. Purity: ≥97% (calc based on dry substance, enzymatic). Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9300 Supplier Enzo Life Sciences Supplier No. ALX480003G001
Alternative name: beta-Nicotinamide Adenine Dinucleotide Phosphate, beta-NADP, Coenzyme II. Purity: ≥97% (calc based on dry substance, enzymatic). Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.