Learn More
Medchemexpress LLC NADP disodium salt | 24292-60-2 | 98.8% | 789.39 | 1 ML
SDP
Catalog No. 5000431826
Encompass_Preferred
NADP disodium salt is a coenzyme involved in cellular electron transfer reactions in biological metabolism. It maintains cellular redox homeostasis, regulates many biological events, and provides reducing ability for synthetic metabolic reactions and redox balance. NADPH, derived from NADP, plays a multifunctional role in regulating inflammation, redox homeostasis, and synthetic metabolism processes.
  • Disodium salt form of NADP.
  • Coenzyme involved in cellular electron transfer reactions.
  • Alternately oxidized (NADP+) and reduced (NADPH).
  • Maintains cellular redox homeostasis.
  • Regulates many biological events, including cellular metabolism.
  • NADPH is a universal electron donor.
  • Provides reducing ability for synthetic metabolic reactions and redox balance.
  • Regulates inflammation, redox homeostasis, and synthetic metabolism processes.
Catalog No. 50-004-31826 Supplier Medchemexpress LLC Supplier No. HYF0002A10MM/1ML
NADP disodium salt is a coenzyme involved in cellular electron transfer reactions in biological metabolism. It maintains cellular redox homeostasis, regulates many biological events, and provides reducing ability for synthetic metabolic reactions and redox balance. NADPH, derived from NADP, plays a multifunctional role in regulating inflammation, redox homeostasis, and synthetic metabolism processes.
  • Disodium salt form of NADP.
  • Coenzyme involved in cellular electron transfer reactions.
  • Alternately oxidized (NADP+) and reduced (NADPH).
  • Maintains cellular redox homeostasis.
  • Regulates many biological events, including cellular metabolism.
  • NADPH is a universal electron donor.
  • Provides reducing ability for synthetic metabolic reactions and redox balance.
  • Regulates inflammation, redox homeostasis, and synthetic metabolism processes.