Learn More
Sigma Aldrich Fine Chemicals Biosciences NADP Disodium salt
Catalog No. 502897222
Encompass
Nicotinamide adenine dinucleotide phosphate (NADP) and NADPH form a redox pair. The NADPH/NADP ratio regulates the intracellular redox potential thereby influencing metabolic reactions in vivo.
Catalog No. 50-289-7222 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10128031001
Nicotinamide adenine dinucleotide phosphate (NADP) and NADPH form a redox pair. The NADPH/NADP ratio regulates the intracellular redox potential thereby influencing metabolic reactions in vivo.