Learn More
TARGETMOL CHEMICALS INC NADP DISODIUM SALT 500MG
SDP
Catalog No. 502580953
Encompass_Preferred
Also available in 100 mg 200 mg and bulk. Please contact Fisher for quotes. NADP disodium salt (Disodium NADP) is a nicotinamide adenine dinucleotide. NADP disodium salt is a key redox cofactor for electron transfer in the metabolism serves as an electron carrier in a number of reactions being alternately oxidized (NADP+) and reduced (NADPH). purity: 98%
Catalog No. 50-258-0953 Supplier TARGETMOL CHEMICALS INC Supplier No. T22446500MG
Also available in 100 mg 200 mg and bulk. Please contact Fisher for quotes. NADP disodium salt (Disodium NADP) is a nicotinamide adenine dinucleotide. NADP disodium salt is a key redox cofactor for electron transfer in the metabolism serves as an electron carrier in a number of reactions being alternately oxidized (NADP+) and reduced (NADPH). purity: 98%