Learn More
Enzo Life Sciences NADP. disodium salt (50mg). CAS: 24292-60-2
SDP
Catalog No. 502009301
Encompass_Preferred
Alternative name: beta-Nicotinamide Adenine Dinucleotide Phosphate, beta-NADP, Coenzyme II. Purity: ≥97% (calc based on dry substance, enzymatic). Appearance: White to light yellow powder. Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9301 Supplier Enzo Life Sciences Supplier No. ALX480003M050
Add to Cart
Add to Cart
Alternative name: beta-Nicotinamide Adenine Dinucleotide Phosphate, beta-NADP, Coenzyme II. Purity: ≥97% (calc based on dry substance, enzymatic). Appearance: White to light yellow powder. Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.