Learn More
Sigma Aldrich Fine Chemicals Biosciences NADP Disodium salt
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 10128040001
Encompass
Nicotinamide adenine dinucleotide phosphate (NADP) and NADPH form a redox pair. The NADPH/NADP ratio regulates the intracellular redox potential thereby influencing metabolic reactions in vivo.
Catalog No. 50-290-3278