Learn More
Cayman Chemical NADP+ (hydrate), Purity: ≥90%, 1 g
SDP
Catalog No. 502981142
Encompass_Preferred
The oxidized form of NADPH; serves as a cofactor in various biological reactions
Catalog No. 50-298-1142 Supplier Cayman Chemical Supplier No. 210451 G
The oxidized form of NADPH; serves as a cofactor in various biological reactions