Learn More
Cayman Chemical NADP hydrate 250mg
SDP
Catalog No. 502982634
Encompass_Preferred
The oxidized form of NADPH; serves as a cofactor in various biological reactions
Catalog No. 50-298-2634 Supplier Cayman Chemical Supplier No. 21045250 MG
The oxidized form of NADPH; serves as a cofactor in various biological reactions