Learn More
Alkali Scientific NADP [β Nicotinamide Adenine Dinucleotide Phosphate, Oxidized Form, Monosodium Salt Trihydrate], 100 Mg
SDP
Catalog No. 5000660813
Encompass_Preferred
NADP (Nicotinamide Adenine Dinucleotide Phosphate, Oxidized Form, Monosodium Salt Trihydrate) is a crucial coenzyme used in biochemical research, particularly in photosynthesis and metabolic pathways. This 100mg quantity is used to study cellular processes like anabolic reactions, lipid synthesis, and DNA repair. NADP is involved in electron transfer in cells and plays an important role in regulating cellular energy and redox status. The monosodium salt form ensures high solubility, making it suitable for various biochemical assays. It is often used in enzyme activity studies and for analyzing oxidative stress in cellular systems.
Catalog No. 50-006-60813 Supplier Alkali Scientific Supplier No. ADP001CP
NADP (Nicotinamide Adenine Dinucleotide Phosphate, Oxidized Form, Monosodium Salt Trihydrate) is a crucial coenzyme used in biochemical research, particularly in photosynthesis and metabolic pathways. This 100mg quantity is used to study cellular processes like anabolic reactions, lipid synthesis, and DNA repair. NADP is involved in electron transfer in cells and plays an important role in regulating cellular energy and redox status. The monosodium salt form ensures high solubility, making it suitable for various biochemical assays. It is often used in enzyme activity studies and for analyzing oxidative stress in cellular systems.