Learn More
Bioworld NADP-SEPAROPOREAGAROSE) 4
Supplier:  Bioworld 201810232
Encompass
NADP-SEPAROPORE (AGAROSE) 4B, LYOPHILIZED: NADP Separopore NADP covalently bound to Separopore has a high affinity for NADP- dependent enzymes and dehydrogenases. Acetyl-CoA binds to NADP Separopore twelve times higher than NAD-Separopo...
Catalog No. 50-197-719