Learn More
Medchemexpress LLC NADP sodium hydrate | 698999-85-8 | 99.0% | 783.40 | 100 MG
SDP
Catalog No. 5000435847
Encompass_Preferred
NADP sodium hydrate is the sodium salt hydrate form of NADP, a coenzyme involved in cellular electron transfer reactions in biological metabolism. It maintains cellular redox homeostasis and regulates many biological events, including cellular metabolism. NADPH, its reduced form, acts as a universal electron donor, providing reducing ability for synthetic metabolic reactions and redox balance.
  • Maintains cellular redox homeostasis.
  • Regulates cellular metabolism.
  • Provides reducing ability for synthetic metabolic reactions.
  • Plays a multifunctional role in regulating inflammation.
  • Important for redox balance.
  • Impairment can affect folate metabolism and nucleotide biosynthesis.
  • Can form NAADP to mobilize Ca2+ and influence cell migration.
  • Upregulation of de novo synthesis can promote breast cancer metastasis.
  • Protects renal cells from oxidative stress.
Catalog No. 50-004-35847 Supplier Medchemexpress LLC Supplier No. HY113325A100MG
NADP sodium hydrate is the sodium salt hydrate form of NADP, a coenzyme involved in cellular electron transfer reactions in biological metabolism. It maintains cellular redox homeostasis and regulates many biological events, including cellular metabolism. NADPH, its reduced form, acts as a universal electron donor, providing reducing ability for synthetic metabolic reactions and redox balance.
  • Maintains cellular redox homeostasis.
  • Regulates cellular metabolism.
  • Provides reducing ability for synthetic metabolic reactions.
  • Plays a multifunctional role in regulating inflammation.
  • Important for redox balance.
  • Impairment can affect folate metabolism and nucleotide biosynthesis.
  • Can form NAADP to mobilize Ca2+ and influence cell migration.
  • Upregulation of de novo synthesis can promote breast cancer metastasis.
  • Protects renal cells from oxidative stress.