Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NADP sodium salt | 1184-16-3 | MFCD00150480 | 98.7% | 765.39 | C21H27N7NaO17P3 | 10 MG
SDP

Catalog No. 5000296988
Encompass_Preferred

NADP sodium salt is the sodium salt form of nicotinamide adenine dinucleotide phosphate supplied as an analytical standard for research and analytical applications. It participates in cellular redox reactions by cycling between oxidized (NADP+) and reduced (NADPH) states, and is used as a reference reagent in biochemical and analytical assays.

  • Purity 98.7% (analytical)
  • Molecular weight 765.39 g/mol
  • Molecular formula C21H27N7NaO17P3
  • Solid, white to off-white appearance
  • Available in small research pack sizes, including 10 MG
  • Intended for research and analytical applications; for research use only

Catalog No. 50-002-96988 Supplier Medchemexpress LLC Supplier No. HYF0002R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NADP sodium salt is the sodium salt form of nicotinamide adenine dinucleotide phosphate supplied as an analytical standard for research and analytical applications. It participates in cellular redox reactions by cycling between oxidized (NADP+) and reduced (NADPH) states, and is used as a reference reagent in biochemical and analytical assays.

  • Purity 98.7% (analytical)
  • Molecular weight 765.39 g/mol
  • Molecular formula C21H27N7NaO17P3
  • Solid, white to off-white appearance
  • Available in small research pack sizes, including 10 MG
  • Intended for research and analytical applications; for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.