Learn More
Cayman Chemical ε-NADP+ (sodium salt), Purity: ≥95%, 3.8 mg
SDP
Catalog No. 502916675
Encompass_Preferred
A fluorescent derivative of NADP+; ex/em = 310/400 nm, respectively; has been used to determine the binding stoichiometry of NADP+ to FASN
Catalog No. 50-291-6675 Supplier Cayman Chemical Supplier No. 383873.8 MG
A fluorescent derivative of NADP+; ex/em = 310/400 nm, respectively; has been used to determine the binding stoichiometry of NADP+ to FASN