Learn More
Cayman Chemical NADP+ (sodium salt hydrate), Purity: ≥90%, CAS: 698999-85-8, 1 g
SDP
Catalog No. 502890634
Encompass_Preferred
The oxidized form of NADPH; serves as a cofactor in various biological reactions
Catalog No. 50-289-0634 Supplier Cayman Chemical Supplier No. 100046751 G
The oxidized form of NADPH; serves as a cofactor in various biological reactions