Learn More
Cayman Chemical NADP+ (sodium salt hydrate), 698999-85-8, 100 mg
SDP
Supplier:  Cayman Chemical 10004675100
Encompass_Preferred
The oxidized form of NADPH; serves as a cofactor in various biological reactions
Catalog No. NC1409341