Learn More
Cayman Chemical NADP+ (sodium salt hydrate), Purity: ≥90%, CAS: 698999-85-8, 500 mg
SDP
Catalog No. 502891187
Encompass_Preferred
The oxidized form of NADPH; serves as a cofactor in various biological reactions
Catalog No. 50-289-1187 Supplier Cayman Chemical Supplier No. 10004675500 MG
The oxidized form of NADPH; serves as a cofactor in various biological reactions