Learn More
Cayman Chemical NADP+ (sodium salt hydrate), Purity: ≥90%, CAS: 698999-85-8, 100 mg
SDP
Catalog No. 502891121
Encompass_Preferred
The oxidized form of NADPH; serves as a cofactor in various biological reactions
Catalog No. 50-289-1121 Supplier Cayman Chemical Supplier No. 10004675100 MG
The oxidized form of NADPH; serves as a cofactor in various biological reactions