Learn More
Research Products International Corp B NADP [B-Nicotinamide Adenine Dinucleotide Phosphate, Reduced Form, Tetra Sodium Salt Trihydrate], 500 Milligrams

Supplier: Research Products International Corp N201400.5
- Substance Name: β-Nicotinamide Adenine Dinucleotide Phosphate, Reduced Form, Tetra Sodium Salt Trihydrate
- CAS Number: 2646-71-1
- Molecular Formula: C21H26N7O17 P3Na4 • 3H2O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.