Learn More
Cayman Chemical NADPH Cytochrome P450 50ug
SDP
Catalog No. 502947681
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged NADPH cytochrome P450 reductase expressed in E. coli • Amino acids: 24-677 • MW: 76.4 kDa
Catalog No. 50-294-7681 Supplier Cayman Chemical Supplier No. 3301550 UG
Source: Active recombinant human C-terminal His-tagged NADPH cytochrome P450 reductase expressed in E. coli • Amino acids: 24-677 • MW: 76.4 kDa