Learn More
Medchemexpress LLC NADPH Oxidase 4 Antibody | 100 UL
SDP
Catalog No. 5000442283
Encompass_Preferred
NADPH Oxidase 4 Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody targeting NADPH Oxidase 4. It reacts with human, mouse, and rat. This antibody is for research use only. The NADPH Oxidase 4 gene encodes a member of the NOX-family of enzymes, acting as an oxygen sensor that catalyzes the reduction of molecular oxygen to reactive oxygen species (ROS). These ROS are involved in signal transduction, cell differentiation, and tumor cell growth.
  • Rabbit-derived, non-conjugated IgG polyclonal antibody
  • Targets NADPH Oxidase 4
  • Reacts with human, mouse, and rat
  • Predicted and observed band size: 67 kDa
  • Affinity purified
  • Liquid appearance
  • Research use only
  • Applications: WB, IHC-P, ICC/IF, IP
  • Stored at -20°C for 1 year; avoid repeated freeze/thaw cycles
Catalog No. 50-004-42283 Supplier Medchemexpress LLC Supplier No. HYP80762100UL
NADPH Oxidase 4 Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody targeting NADPH Oxidase 4. It reacts with human, mouse, and rat. This antibody is for research use only. The NADPH Oxidase 4 gene encodes a member of the NOX-family of enzymes, acting as an oxygen sensor that catalyzes the reduction of molecular oxygen to reactive oxygen species (ROS). These ROS are involved in signal transduction, cell differentiation, and tumor cell growth.
  • Rabbit-derived, non-conjugated IgG polyclonal antibody
  • Targets NADPH Oxidase 4
  • Reacts with human, mouse, and rat
  • Predicted and observed band size: 67 kDa
  • Affinity purified
  • Liquid appearance
  • Research use only
  • Applications: WB, IHC-P, ICC/IF, IP
  • Stored at -20°C for 1 year; avoid repeated freeze/thaw cycles