Learn More
MOLTOX Molecular Toxicology Inc 153 mg of crystalline NADP
SDP
Catalog No. 50948934
Encompass
NADPH REGENSYS B, 50 mL size refers to final volume of S-9 mix per pkg, 153 mg of crystalline NADP, Add to A, pkg w/ dessicant, Store in freezer
Catalog No. 50-948-934 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 60201.5L
Add to Cart
Add to Cart
NADPH REGENSYS B, 50 mL size refers to final volume of S-9 mix per pkg, 153 mg of crystalline NADP, Add to A, pkg w/ dessicant, Store in freezer