Learn More
Cayman Chemical NADPH (sodium salt), 2646-71-1, 50 mg
SDP
Supplier:  Cayman Chemical 900074350
Encompass_Preferred
The reduced form of the electron acceptor NADP+ and acts as an electron donor in various biological reactions
Catalog No. NC1046234