Learn More
Selleck Chemical LLC NADPH tetrasodium salt-250MG
SDP
Catalog No. 503136828
Encompass_Preferred
NADPH tetrasodium salt is a ubiquitous coenzyme that acts as an electron donor in many reactions utilizing dehydrogenase and reductase enzymes It plays a crucial role as a cofactor in the biosynthesis of nucleotides proteins and fatty acids It is essential for neutralizing elevated levels of reactive oxygen species (ROS) generated by increased metabolic activity
Catalog No. 50-313-6828 Supplier Selleck Chemical LLC Supplier No. E1689250MG
NADPH tetrasodium salt is a ubiquitous coenzyme that acts as an electron donor in many reactions utilizing dehydrogenase and reductase enzymes It plays a crucial role as a cofactor in the biosynthesis of nucleotides proteins and fatty acids It is essential for neutralizing elevated levels of reactive oxygen species (ROS) generated by increased metabolic activity