Learn More
Medchemexpress LLC NADPH (tetrasodium salt) | 2646-71-1 | 99.99% | 833.35 | 50 MG
SDP
Catalog No. 501874599
Encompass_Preferred
NADPH tetrasodium salt functions as an important cofactor in a variety of metabolic and biosynthetic pathways. It is an endogenous inhibitor of ferroptosis. NADPH tetrasodium salt plays a vital role in the biosynthesis of agents, chiral alcohols, fatty acids, and biopolymers, while also being required for lipid biosynthesis, biomass formation, and cell replication.
  • Functions as an important cofactor in various metabolic and biosynthetic pathways.
  • Is an endogenous inhibitor of ferroptosis.
  • Plays a vital role in the biosynthesis of agents, chiral alcohols, fatty acids, and biopolymers.
  • Required for lipid biosynthesis, biomass formation, and cell replication.
Catalog No. 50-187-4599 Supplier Medchemexpress LLC Supplier No. HYF000350MG
NADPH tetrasodium salt functions as an important cofactor in a variety of metabolic and biosynthetic pathways. It is an endogenous inhibitor of ferroptosis. NADPH tetrasodium salt plays a vital role in the biosynthesis of agents, chiral alcohols, fatty acids, and biopolymers, while also being required for lipid biosynthesis, biomass formation, and cell replication.
  • Functions as an important cofactor in various metabolic and biosynthetic pathways.
  • Is an endogenous inhibitor of ferroptosis.
  • Plays a vital role in the biosynthesis of agents, chiral alcohols, fatty acids, and biopolymers.
  • Required for lipid biosynthesis, biomass formation, and cell replication.