Learn More
Cayman Chemical NADPH (cyclohexyl ammonium salt), Purity: ≥95%, CAS: 100929-71-3, 10 mg
SDP
Catalog No. 502981713
Encompass_Preferred
The reduced form of the electron acceptor NADP+ and acts as an electron donor in various biological reactions
Catalog No. 50-298-1713 Supplier Cayman Chemical Supplier No. 2094510 MG
The reduced form of the electron acceptor NADP+ and acts as an electron donor in various biological reactions