Learn More
Cayman Chemical NADPHcyclohexyl amonIM sa 50mg
SDP
Catalog No. 502983876
Encompass_Preferred
The reduced form of the electron acceptor NADP+ and acts as an electron donor in various biological reactions
Catalog No. 50-298-3876 Supplier Cayman Chemical Supplier No. 2094550 MG
The reduced form of the electron acceptor NADP+ and acts as an electron donor in various biological reactions